missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human XCL1 Protein
A DNA sequence encoding the Homo sapiens (Human) XCL1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8667-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002986.1 | |
XCL1 Protein | |
Research Use Only | |
XCL1 | |
Recombinant Protein | |
E. coli | |
VGSEVSDKRT CVSLTTQRLP VSRIKTYTIT EGSLRAVIFI TKRGLKVCAD PQATWVRDVV RSMDRKSNTR NNMIQTKPTG TQQSTNTAVT LTG | |
Greater than 90% as determined by SDS-PAGE. |
6375 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |