missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human XAGE1A Protein

A cDNA sequence encoding the XAGE1A was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13970-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15925279

  • 160.00€ / 5 microgramos


Fecha de envío estimada
del proveedor 09-04-2019
Añadir a la cesta

Especificación

Especificación

XAGE1A Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
XAGE1A protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 20% glycerol and 2mM DTT.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMESPKKK NQQLKVGILH LGSRQKKIRI QLRSQCATWK VICKSCISQT PGINLDLGSG VKVKIIPKEE HCKMPEAGEE QPQV
Videos