missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Xaa-Pro aminopeptidase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Xaa-Pro aminopeptidase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8152-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001161076.1 | |
Xaa-Pro aminopeptidase 1 Protein | |
Research Use Only | |
XPNPEP1 | |
Recombinant Protein | |
E. coli | |
PPKVTSELLR QLRQAMRNSE YVTEPIQAYI IPSGDAHQSE YIAPCDCRRA FVSGFDGSAG TAIITEEHAA MWTDGRYFLQ AAKQMDSNWT LMKMGLKDTP TQEDWLVSVL PEGSRVGVDP LIIPTDYWKK MAKVLRSAGH HLIPVKENLV DKIWTDRPER PCKPLLTLGL DYTGISWKDK VADLRLKMAE RNVMWFVVTA LDEIAWLFNL RGSDVEHNPV FFSYAIIGLE TIMLFIDGDR IDAPSVKEHL LLDLGLEAEY RIQVHPYKSI LSELKALCAD LSPREKVWVS DKASYAVSET IPKDHRCCMP YTPICIAKAV KNSAESEGMR RAHIKDAVAL CELFNWLEKE VPKGGVTEIS AADKAEEFRR QQADFVDLSF PTISSTGPNG AIIHYAPVPE TNRTLSLDEV YLIDSGAQYK DGTTDVTRTM HFGTPTAYEK ECFTYVLKGH IAVSAAVFPT GTKGHLLDSF ARSALWDSGL DYLHGTGHGV GSFLNVHEGP CGISYKTFSD EPLEAGMIVT DEPGYYEDGA FGIRIENVVL VVPVKTKYNF NNRGSLTFEP LTLVPIQTKM IDVDSLTDKE CDWLNNYHLT CRDVIGKELQ KQGRQEALEW LIRETQPISK QH | |
Greater than 90% as determined by SDS-PAGE. |
7511 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |