missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human X-ray repair cross-complementing protein 1
A DNA sequence encoding the Homo sapiens (Human) X-ray repair cross-complementing protein 6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8378-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001275905.1 | |
X-ray repair cross-complementing protein 1 | |
Research Use Only | |
XRCC6 | |
Recombinant Protein | |
E. coli | |
SYYKTEGDEE AEEEQEENLE ASGDYKYSGR DSLIFLVDAS KAMFESQSED ELTPFDMSIQ CIQSVYISKI ISSDRDLLAV VFYGTEKDKN SVNFKNIYVL QELDNPGAKR ILELDQFKGQ QGQKRFQDMM GHGSDYSLSE VLWVCANLFS DVQFKMSHKR IMLFTNEDNP HGNDSAKASR ARTKAGDLRD TGIFLDLMHL KKPGGFDISL FYRDIIS | |
Greater than 80% as determined by SDS-PAGE. |
2547 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |