missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VRK3 Protein

A cDNA sequence encoding the VRK3 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13947-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15935209

  • 107.00€ / 2 microgramos


Fecha estimada de envío
25-06-2020
Añadir a la cesta

Especificaciones

Especificaciones

VRK3 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The VRK3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 40% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMISFCPD CGKSIQAAFK FCPYCGNSLP VEEHVGSQTF VNPHVSSFQG SKRGLNSSFE TSPKKVKWSS TVTSPRLSLF SDGDSSESED TLSSSERSKG SGSRPPTPKS SPQKTRKSPQ VTRGSPQKTS CSPQKTRQSP QTLKRSRVTT SLEALPTGTV LTDKSGRQWK LKSFQTRDNQ GILYEAAPTS TLTCDSGPQK QKFSLKLDAK DGRLFNEQNF FQRAAKPLQV NKWKKLYSTP LLAIPTCMGF GVHQDKYRFL VLPSLGRSLQ SALDVSPKHV LSERSVLQVA CRLLDALEFL HENEYVHGNV TAENIFVDPE DQSQVTLAGY GFAFRYCPSG KHVAYVEGSR SPHEGDLEFI SMDLHKGCGP SRRSDLQSLG YCMLKWLYGF LPWTNCLPNT EDIMKQKQKF VDKPGPFVGP CGHWIRPSET LQKYLKVVMA LTYEEKPPYA MLRNNLEALL QDLRVSPYDP IGLPMVP
Videos
Documentos