missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UROD Protein

A cDNA sequence encoding the UROD was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13907-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915089

  • 2583.00€ / 1 miligramo


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

7389
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
UROD
Human
His
UROD Human solution containing 20mM Tris pH 8.0, 1mM DTT, 0.1M NaCl, 1mM EDTA and 20% glycerol.
UROD Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MEANGLGPQG FPELKNDTFL RAAWGEETDY TPVWCMRQAG RYLPEFRETR AAQDFFSTCR SPEACCELTL QPLRRFPLDA AIIFSDILVV PQALGMEVTM VPGKGPSFPE PLREEQDLER LRDPEVVASE LGYVFQAITL TRQRLAGRVP LIGFAGAPWT LMTYMVEGGG SSTMAQAKRW LYQRPQASHQ LLRILTDALV PYLVGQVVAG AQALQLFESH AGHLGPQLFN KFALPYIRDV AKQVKARLRE AGLAPVPMII FAKDGHFALE ELAQAGYEVV GLDWTVAPKK ARECVGKTVT LQVNLDPCAL YASEEEIGQL VKQMLDDFGP HRYIANLGHG LYPDMDPEHV GAFVDAVHKH SRLLRQN
Videos