missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UQCRC2 Protein
A cDNA sequence encoding the UQCRC2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13904-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
UQCRC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSLKVAPKVK ATAAPAGAPP QPQDLEFTKL PNGLVIASLE NYSPVSRIGL FIKAGSRYED FSNLGTTHLL RLTSSLTTKG ASSFKITRGI EAVGGKLSVT ATRENMAYTV ECLRGDVDIL MEFLLNVTTA PEFRRWEVAD LQPQLKIDKA VAFQNPQTHV IENLHAAAYR NALANPLYCP DYRIGKVTSE ELHYFVQNHF TSARMALIGL GVSHPVLKQV AEQFLNMRGG LGLSGAKANY RGGEIREQNG DSLVHAAFVA ESAVAGSAEA NAFSVLQHVL GAGPHVKRGS NTTSHLHQAV AKATQQPFDV SAFNASYSDS GLFGIYTISQ ATAAGDVIKA AYNQVKTIAQ GNLSNTDVQA AKNKLKAGYL MSVESSECFL EEVGSQALVA GSYMPPSTVL QQIDSVANAD IINAAKKFVS GQKSMAASGN LGHTPFVDEL | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UQCRC2 solution contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea, 0.1M NaCl and 10% glycerol. |