missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UPP1 Protein

A cDNA sequence encoding the UPP1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13900-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915069

  • 5941.00€ / 1 miligramo


Fecha estimada de envío
17-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

UPP1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
UPP1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.2M NaCl and 40% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAATGANAEK AESHNDCPVR LLNPNIAKMK EDILYHFNLT TSRHNFPALF GDVKFVCVGG SPSRMKAFIR CVGAELGLDC PGRDYPNICA GTDRYAMYKV GPVLSVSHGM GIPSISIMLH ELIKLLYYAR CSNVTIIRIG TSGGIGLEPG TVVITEQAVD TCFKAEFEQI VLGKRVIRKT DLNKKLVQEL LLCSAELSEF TTVVGNTMCT LDFYEGQGRL DGALCSYTEK DKQAYLEAAY AAGVRNIEME SSVFAAMCSA CGLQAAVVCV TLLNRLEGDQ ISSPRNVLSE YQQRPQRLVS YFIKKKLSKA
Videos