missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UPF0454 protein C12orf49 Protein

A DNA sequence encoding the Homo sapiens (Human) UPF0454 protein C12orf49, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8088-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052601

  • 253.00€ / 50 microgramos


Fecha estimada de envío
22-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_079014.1
UPF0454 protein C12orf49 Protein
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
79794
Greater than 90% as determined by SDS-PAGE.
Research Use Only
C12orf49
Recombinant Protein
E. coli
TFKQEERAVR DRNLLQVHDH NQPIPWKVQF NLGNSSRPSN QCRNSIQGKH LITDELGYVC ERKDLLVNGC CNVNVPSTKQ YCCDGCWPNG CCSAYEYCVS CCLQPNKQLL LERFLNRAAV AFQNLFMAVE DHFELCLAKC RTSSQSVQHE NTYRDPIAKY CYGESPPELF PA
Videos
Documentos