missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBL4A Protein
A cDNA sequence encoding the UBL4A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13873-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
UBL4A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MQLTVKALQG RECSLQVPED ELVSTLKQLV SEKLNVPVRQ QRLLFKGKAL ADGKRLSDYS IGPNSKLNLV VKPLEKVLLE EGEAQRLADS PPPQVWQLIS KVLARHFSAA DASRVLEQLQ RDYERSLSRL TLDDIERLAS RFLHPEVTET MEKGFSKLEH HHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UBL4A protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 100mM NaCl. |