missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ubiquitin-like-conjugating enzyme ATG10 Protein

A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-like-conjugating enzyme ATG10, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8050-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052371

  • 675.00€ / 200 microgramos


Fecha estimada de envío
19-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001124500.1
Ubiquitin-like-conjugating enzyme ATG10 Protein
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
83734
Greater than 80% as determined by SDS-PAGE.
Research Use Only
ATG10
Recombinant Protein
E. coli
MEEDEFIGEK TFQRYCAEFI KHSQQIGDSW EWRPSKDCSD GYMCKIHFQI KNGSVMSHLG ASTHGQTCLP MEEAFELPLD DCEVIETAAA SEVIKYEYHV LYSCSYQVPV LYFRASFLDG RPLTLKDIWE GVHECYKMRL LQGPWDTITQ QEHPILGQPF FVLHPCKTNE FMTPVLKNSQ KINKNVNYIT SWLSIVGPVV GLNLPLSYAK ATSQDERNVP
Videos
Documentos