missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin G76A Protein
A cDNA sequence encoding the Ubiquitin G76A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13870-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
7311; 7314; 6233; 7316 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
Diluted in PBS plus 5% glycerol. |
Ubiquitin G76A Protein | |
Research Use Only | |
RPS27A; UBB; UBC; UBA52 | |
Human | |
Untagged | |
Greater than 95% as determined by SDS-PAGE. |