missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin D Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin D, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6862-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006389.2 | |
Ubiquitin D Protein | |
Research Use Only | |
UBD | |
Recombinant Protein | |
E. coli | |
MAPNASCLCV HVRSEEWDLM TFDANPYDSV KKIKEHVRSK TKVPVQDQVL LLGSKILKPR RSLSSYGIDK EKTIHLTLKV VKPSDEELPL FLVESGDEAK RHLLQVRRSS SVAQVKAMIE TKTGIIPETQ IVTCNGKRLE DGKMMADYGI RKGNLLFLAC YCIGG | |
Greater than 90% as determined by SDS-PAGE. |
10537 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |