missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UBE2E1 Protein

A cDNA sequence encoding the UBE2E1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13857-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914929

  • 103.00€ / 5 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

UBE2E1 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The UBE2E1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 20% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSDDDSR ASTSSSSSSS SNQQTEKETN TPKKKESKVS MSKNSKLLST SAKRIQKELA DITLDPPPNC SAGPKGDNIY EWRSTILGPP GSVYEGGVFF LDITFTPEYP FKPPKVTFRT RIYHCNINSQ GVICLDILKD NWSPALTISK VLLSICSLLT DCNPADPLVG SIATQYMTNR AEHDRMARQW TKRYAT
Videos