missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Type-1 angiotensin II receptor Protein

A DNA sequence encoding the Homo sapiens (Human) Type-1 angiotensin II receptor, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8960-ye-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16057311

  • 2060.00€ / 500 microgramos


Fecha estimada de envío
20-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000676.1
Type-1 angiotensin II receptor Protein
500 μg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
185
Greater than 90% as determined by SDS-PAGE.
Research Use Only
AGTR1
Recombinant Protein
Yeast
LNPLFYGFLG KKFKRYFLQL LKYIPPKAKS HSNLSTKMST LSYRPSDNVS SSTKKPAPCF EVE
Videos
Documentos