missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TSSK2 Protein

A cDNA sequence encoding the TSSK2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13832-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15934849

  • 161.00€ / 10 microgramos


Fecha estimada de envío
17-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

23617
Greater than 85% as determined by SDS-PAGE.
Research Use Only
TSSK2
Human
His
TSSK2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
TSSK2 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDDATVL RKKGYIVGIN LGKGSYAKVK SAYSERLKFN VAVKIIDRKK TPTDFVERFL PREMDILATV NHGSIIKTYE IFETSDGRIY IIMELGVQGD LLEFIKCQGA LHEDVARKMF RQLSSAVKYC HDLDIVHRDL KCENLLLDKD FNIKLSDFGF SKRCLRDSNG RIILSKTFCG SAAYAAPEVL QSIPYQPKVY DIWSLGVILY IMVCGSMPYD DSDIRKMLRI QKEHRVDFPR SKNLTCECKD LIYRMLQPDV SQRLHIDEIL SHSWLQPPKP KATSSASFKR EGEGKYRAEC KLDTKTGLRP DHRPDHKLGA KTQHRLLVVP ENENRMEDRL AETSRAKDHH ISGAEVGKAS T
Videos
Documentos