missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TSN Protein

A cDNA sequence encoding the TSN was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13828-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914839

  • 160.00€ / 20 microgramos


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

TSN Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The TSN protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 100mM NaCl, and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MSVSEIFVEL QGFLAAEQDI REEIRKVVQS LEQTAREILT LLQGVHQGAG FQDIPKRCLK AREHFGTVKT HLTSLKTKFP AEQYYRFHEH WRFVLQRLVF LAAFVVYLET ETLVTREAVT EILGIEPDRE KGFHLDVEDY LSGVLILASE LSRLSVNSVT AGDYSRPLHI STFINELDSG FRLLNLKNDS LRKRYDGLKY DVKKVEEVVY DLSIRGFNKE TAAACVEK
Videos