missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSH Protein
A cDNA sequence encoding the TSH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10901-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TSH Protein | |
Research Use Only | |
11.72 IU/mg vial by Centaur CP. Siemens Centaur CP is standardized against WHO 3rd IRP 81/565. | |
Human | |
Untagged | |
Greater than 95% as determined by SDS-PAGE. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human | |
Lyophilized from a concentrated solution containing 50mM ammonium bicarbonate. |