missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSG101 Protein
A cDNA sequence encoding the TSG101 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13826-1mg
Detalles adicionales : Peso : 0.01000kg
Specifications
TSG101 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMAVS ESQLKKMVSK YKYRDLTVRE TVNVITLYKD LKPVLDSYVF NDGSSRELMN LTGTIPVPYR GNTYNIPICL WLLDTYPYNP PICFVKPTSS MTIKTGKHVD ANGKIYLPYL HEWKHPQSDL LGLIQVMIVV FGDEPPVFSR P | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TSG101 protein solution (0.5 mg/ml) containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |