missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tryptophan 5-hydroxylase 2 Protein

A DNA sequence encoding the Homo sapiens(Human) Tryptophan 5-hydroxylase 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10004-iv-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922738

Cantidad Precio
1 845.00€ / 50 microgramos
5 788.98€ / 50 microgramos
Fecha estimada de envío
04-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_775489.2
Tryptophan 5-hydroxylase 2 Protein
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
121278
Greater than 85% as determined by SDS-PAGE.
Research Use Only
TPH2
Recombinant Protein
Invitro-E. coli
MQPAMMMFSS KYWARRGFSL DSAVPEEHQL LGSSTLNKPN SGKNDDKGNK GSSKREAATE SGKTAVVFSL KNEVGGLVKA LRLFQEKRVN MVHIESRKSR RRSSEVEIFV DCECGKTEFN ELIQLLKFQT TIVTLNPPEN IWTEEEELED VPWFPRKISE LDKCSHRVLM YGSELDADHP GFKDNVYRQR RKYFVDVAMG YKYGQPIPRV EYTEEETKTW GVVFRELSKL YPTHACREYL KNFPLLTKYC GYREDNVPQL EDVSMFLKER SGFTVRPVAG YLSPRDFLAG LAYRVFHCTQ YIRHGSDPLY TPEPDTCHEL LGHVPLLADP KFAQFSQEIG LASLGASDED VQKLATCYFF TIEFGLCKQE GQLRAYGAGL LSSIGELKHA LSDKACVKAF DPKTTCLQEC LITTFQEAYF VSESFEEAKE KMRDFAKSIT RPFSVYFNPY TQSIEILKDT RSIENVVQDL RSDLNTVCDA LNKMNQYLGI
Videos
Documentos