missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tryptophan 5-hydroxylase 2 Protein

A DNA sequence encoding the Homo sapiens(Human) Tryptophan 5-hydroxylase 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10004-iv-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15912738

  • 910.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

NP_775489.2
Tryptophan 5-hydroxylase 2 Protein
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
121278
Greater than 85% as determined by SDS-PAGE.
Research Use Only
TPH2
Recombinant Protein
Invitro-E. coli
MQPAMMMFSS KYWARRGFSL DSAVPEEHQL LGSSTLNKPN SGKNDDKGNK GSSKREAATE SGKTAVVFSL KNEVGGLVKA LRLFQEKRVN MVHIESRKSR RRSSEVEIFV DCECGKTEFN ELIQLLKFQT TIVTLNPPEN IWTEEEELED VPWFPRKISE LDKCSHRVLM YGSELDADHP GFKDNVYRQR RKYFVDVAMG YKYGQPIPRV EYTEEETKTW GVVFRELSKL YPTHACREYL KNFPLLTKYC GYREDNVPQL EDVSMFLKER SGFTVRPVAG YLSPRDFLAG LAYRVFHCTQ YIRHGSDPLY TPEPDTCHEL LGHVPLLADP KFAQFSQEIG LASLGASDED VQKLATCYFF TIEFGLCKQE GQLRAYGAGL LSSIGELKHA LSDKACVKAF DPKTTCLQEC LITTFQEAYF VSESFEEAKE KMRDFAKSIT RPFSVYFNPY TQSIEILKDT RSIENVVQDL RSDLNTVCDA LNKMNQYLGI
Videos