missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TRAPPC4 Protein
A cDNA sequence encoding the TRAPPC4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13806-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
51399 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAIFSVY VVNKAGGLIY QLDSYAPRAE AEKTFSYPLD LLLKLHDERV LVAFGQRDGI RVGHAVLAIN GMDVNGRYTA DGKEVLEYLG NPANYPVSIR FGRPRLTSNE KLMLASMFHS LFAIGSQLSP EQGSSGIEML ETDTFKLHCY QTLTGIKFVV LADPRQAGID SLLRKIYEIY SDFALKNPFY SLEMPIRCEL FDQNLKLALE VAEKAGTFGP GS | |
Greater than 90.0% as determined by SDS-PAGE. |
TRAPPC4 Protein | |
Research Use Only | |
TRAPPC4 | |
Human | |
His | |
TRAPPC4 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 20% glycerol and 1mM DTT. |