missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcriptional regulator ERG Protein

A DNA sequence encoding the Homo sapiens (Human) Transcriptional regulator ERG, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5996-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919229

  • 1141.00€ / 500 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001129626.1
Transcriptional regulator ERG Protein
500 μg
Not determined.
Full Length of Isoform 5
Human
His-SUMO
Tris-based buffer, 50% glycerol
2078
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ERG
Recombinant Protein
E. coli
MIQTVPDPAA HIKEALSVVS EDQSLFECAY GTPHLAKTEM TASSSSDYGQ TSKMSPRVPQ QDWLSQPPAR VTIKMECNPS QVNGSRNSPD ECSVAKGGKM VGSPDTVGMN YGSYMEEKHM PPPNMTTNER RVIVPADPTL WSTDHVRQWL EWAVKEYGLP DVNILLFQNI DGKELCKMTK DDFQRLTPSY NADILLSHLH YLRETPLPHL TSDDVDKALQ NSPRLMHARN TGGAAFIFPN TSVYPEATQR ITTRPDLPYE PPRRSAWTGH GHPTPQSKAA QPSPSTVPKT EDQRPQLDPY QILGPTSSRL ANPGWTQ
Videos