missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-4 Protein

A DNA sequence encoding the Homo sapiens (Human) Transcriptional enhancer factor TEF-4, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7533-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16059220

  • 1857.00€ / 1 miligramo


Fecha estimada de envío
26-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001243587.1
Transcriptional enhancer factor TEF-4 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
8463
Greater than 80% as determined by SDS-PAGE.
Research Use Only
TEAD2
Recombinant Protein
E. coli
MGEPRAGAAL DDGSGWTGSE EGSEEGTGGS EGAGGDGGPD AEGVWSPDIE QSFQEALAIY PPCGRRKIIL SDEGKMYGRN ELIARYIKLR TGKTRTRKQV SSHIQVLARR KSREIQSKLK DQVSKDKAFQ TMATMSSAQL ISAPSLQAKL GPTGPQASEL FQFWSGGSGP PWNVPDVKPF SQTPFTLSLT PPSTDLPGYE PPQALSPLPP PTPSPPAWQA RGLGTARLQL VEFSAFVEPP DAVDSYQRHL FVHISQHCPS PGAPPLESVD VRQIYDKFPE KKGGLRELYD RGPPHAFFLV KFWADLNWGP SGEEAGAGGS ISSGGFYGVS SQYESLEHMT LTCSSKVCSF GKQVVEKVET ERAQLEDGRF VYRLLRSPMC EYLVNFLHKL RQLPERYMMN SVLENFTILQ VVTNRDTQEL LLCTAYVFEV STSERGAQHH IYRLVRD
Videos
Documentos