missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TPM1 Protein
A cDNA sequence encoding the TPM1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13794-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TPM1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDAIKKKMQM LKLDKENALD RAEQAEADKK AAEDRSKQLE DELVSLQKKL KGTEDELDKY SEALKDAQEKLELAEKKATD AEADVASLNR RIQLVEEELD RAQERLATAL QKLEEAEKAA DESERGMKVI ESRAQKDEEK MEIQEIQLKE AKHIAEDADRKYEEVARKLV IIESDLERAE ERAELSEGQV RQLEEQLRIM DQTLKALMAA EDKYSQKEDR YEEEIKVLSD KLKEAETRAE FAERSVTKLEKSIDDLEDEL YAQKLKYKAI SEELDHALND MTSM | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TPM1 0.5 mg/ml protein solution contains 20mM Tris-HCl buffer pH 8.0, 1mM DTT, 0.1M NaCl and 20% glycerol. |