missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human tPA / PLAT Protein

A DNA sequence encoding the Homo sapiens (Human) tPA / PLAT, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6501-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053760

  • 160.00€ / 10 microgramos


Fecha estimada de envío
24-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000921.1
tPA / PLAT Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
5327
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PLAT
Recombinant Protein
E. coli
SYQVICRDEK TQMIYQQHQS WLRPVLRSNR VEYCWCNSGR AQCHSVPVKS CSEPRCFNGG TCQQALYFSD FVCQCPEGFA GKCCEIDTRA TCYEDQGISY RGTWSTAESG AECTNWNSSA LAQKPYSGRR PDAIRLGLGN HNYCRNPDRD SKPWCYVFKA GKYSSEFCST PACSEGNSDC YFGNGSAYRG THSLTESGAS CLPWNSMILI GKVYTAQNPS AQALGLGKHN YCRNPDGDAK PWCHVLKNRR LTWEYCDVPS CSTCGLRQYS QPQFRIKGGL FADIASHPWQ AAIFAKHRRS PGERFLCGGI LISSCWILSA AHCFQERFPP HHLTVILGRT YRVVPGEEEQ KFEVEKYIVH KEFDDDTYDN DIALLQLKSD SSRCAQESSV VRTVCLPPAD LQLPDWTECE LSGYGKHEAL SPFYSERLKE AHVRLYPSSR CTSQHLLNRT VTDNMLCAGD TRSGGPQANL HDACQGDSGG PLVCLNDGRM TLVGIISWGL GCGQKDVPGV YTKVTNYLDW I
Videos
Documentos