missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNNI2 Protein
A cDNA sequence encoding the TNNI2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13776-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
7136 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGDEEKRNRA ITARRQHLKS VMLQIAATEL EKEESRREAE KQNYLAEHCP PLHIPGSMSE VQELCKQLHA KIDAAEEEKY DMEVRVQKTS KELEDMNQKL FDLRGKFKRP PLRRVRMSAD AMLKALLGSK HKVCMDLRAN LKQVKKEDTE KERDLRDVGD WRKNIEEKSG MEGRKKMFES ES | |
Greater than 90% as determined by SDS-PAGE. |
TNNI2 Protein | |
Research Use Only | |
TNNI2 | |
Human | |
Untagged | |
The TNNI2 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea, 20% glycerol and 0.2M NaCl. |