missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNFRSF13B / TACI / CD267 Protein

A DNA sequence encoding the Human TNFRSF13B / TACI / CD267, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10368-ec-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923648

  • 114.00€ / 5 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_036584.1
TNFRSF13B / TACI / CD267 Protein
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MSGLGRSRRG GRSRVDQEER FPQGLWTGVA MRSCPEEQYW DPLLGTCMSC KTICNHQSQR TCAAFCRSLS CRKEQGKFYD HLLRDCISCA SICGQHPKQC AYFCENKLRS PVNLPPELRR QRSGEVENNS DNSGRYQGLE HRGSEASPAL PGLKLSADQV
23495
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
TNFRSF13B
The biological activity is determined by its ability to block human BAFF induced T2B cell survival using a concentration range of 1.0-3.0 ug/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos