missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF11A Protein
A DNA sequence encoding the Homo sapiens (Human) TNFRSF11A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8604-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001257878.1 | |
TNFRSF11A Protein | |
Research Use Only | |
TNFRSF11A | |
Recombinant Protein | |
E. coli | |
LQIAPPCTSE KHYEHLGRCC NKCEPGKYMS SKCTTTSDSV CLPCGPDEYL DSWNEEDKCL LHKVCDTGKA LVAVVAGNST TPRRCACTAG YHWSQDCECC RRNTECAPGL GAQHPLQLNK DTVCKPCLAG YFSDAFSSTD KCRPWTNCTF LGKRVEHHGT EKSDAVCSSS LPARK | |
Greater than 90% as determined by SDS-PAGE. |
8792 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |