missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF10D Protein
A cDNA sequence encoding the TNFRSF10D was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10888-1ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TNFRSF10D Protein | |
Research Use Only | |
Measured in a neutralizing assay using Jurkat human T lymphocyte. The ED50 for this effect is less or equal to 10 ng/ml in the presence of 2ng/ml TRAIL. | |
Human | |
His | |
TNFRSF10D protein solution (0.5 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |
1 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
ATIPRQDEVP QQTVAPQQQR RSLKEEECPA GSHRSEYTGA CNPCTEGVDY TIASNNLPSC LLCTVCKSGQ TNKSSCTTTR DTVCQCEKGS FQDKNSPEMC RTCRTGCPRG MVKVSNCTPR SDIKCKNESA ASSTGKTPAA EETVTTILGM LASPYHVEPK SCDKTHTCPP CPAPELLGGPSVFLFPPKPK DTLMISRTPE VTCVVVDVSH EDPEVKFNWY VDGVEVHNAK TKPREEQYNS TYRVVSVLTV LHQDWLNGKE YKCKVSNKAL PAPIEKTISK AKGQPREPQV YTLPPSRDEL TKNQVSLTCL VKGFYPSDIA VEWESNGQPE NNYKTTPPVL DSDGSFFLYS KLTVDKSRWQQGNVFSCSVM HEALHNHYTQ KSLSLSPGKH HHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |