missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TIFA Protein

A cDNA sequence encoding the TIFA was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13752-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914589

  • 5941.00€ / 1 miligramo


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

TIFA Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TIFA protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMTSFED ADTEETVTCL QMTVYHPGQL QCGIFQSISF NREKLPSSEV VKFGRNSNIC HYTFQDKQVS RVQFSLQLFK KFNSSVLSFE IKNMSKKTNL IVDSRELGYL NKMDLPYRCM VRFGEYQFLM EKEDGESLEF FETQFILSPR SLLQENNWPP HRPIPEYGTY SLCSSQSSSP TEMDENES
Videos