missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thymosin beta-4 Protein
A DNA sequence encoding the Human Thymosin beta-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10182-ec-20ug
Additional Details : Peso : 0.01000kg
Specifications
NP_066932.1 | |
Thymosin beta-4 Protein | |
Research Use Only | |
TMSB4X | |
The biological activity determined by its ability to produce a protective effect against hydrogen peroxide in primary lung fibroblasts is in a concentration range of 0.5 - 10 ug/ml. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
7114 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SDKPDMAEIE KFDKSKLKKT ETQEKNPLPS KETIEQEKQA GES | |
Greater than 97% as determined by SDS-PAGE and HPLC. |