missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human THG1L Protein

A cDNA sequence encoding the THG1L was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13742-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914559

  • 160.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 03-04-2019
Añadir a la cesta

Especificación

Especificación

54974
Greater than 90% as determined by SDS-PAGE.
Research Use Only
THG1L
Human
His
The THG1L solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol.
THG1L Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAKSKFE YVRDFEADDT CLAHCWVVVR LDGRNFHRFA EKHNFAKPND SRALQLMTKC AQTVMEELED IVIAYGQSDE YSFVFKRKTN WFKRRASKFM THVASQFASS YVFYWRDYFE DQPLLYPPGF DGRVVVYPSN QTLKDYLSWR QADCHINNLY NTVFWALIQQ SGLTPVQAQG RLQGTLAADK NEILFSEFNI NYNNELPMYR KGTVLIWQKV DEVMTKEIKL PTEMEGKKMA VTRTRTKPVP LHCDIIGDAF WKEHPEILDE DS
Videos