missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TEN1 Protein
A cDNA sequence encoding the TEN1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13722-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TEN1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMMLPKPG TYYLPWEVSA GQVPDGSTLR TFGRLCLYDM IQSRVTLMAQ HGSDQHQVLV CTKLVEPFHA QVGSLYIVLG ELQHQQDRGS VVKARVLTCV EGMNLPLLEQ AIREQRLYKQ ERGGSQ | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TEN1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.15M NaCl, 10% glycerol and 1mM DTT. |