missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TBC1D22B Protein

A cDNA sequence encoding the TBC1D22B was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13694-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15924409

  • 160.00€ / 5 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

TBC1D22B Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TBC1D22B protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4) 30% glycerol and 1mM DTT.
Greater than 80% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAAENSK QFWKRSAKLP GSIQPVYGAQ HPPLDPRLTK NFIKERSKVN TVPLKNKKAS SFHEFARNTS DAWDIGDDEE EDFSSPSFQT LNSKVALATA AQVLENHSKL RVKPERSQST TSDVPANYKV IKSSSDAQLS RNSSDTCLRN PLHKQQSLPL RPIIPLVARI SDQNASGAPP MTVREKTRLE KFRQLLSSQN TDLDELRKCS WPGVPREVRP ITWRLLSGYL PANTERRKLT LQRKREEYFG FIEQYYDSRN EEHHQDTYRQ IHIDIPRTNP LIPLFQQPLV QEIFERILFI WAIRHPASGY VQGINDLVTP FFVVFLSEYV EEDVENFDVT NLSQDMLRSI EADSFWCMSK LLDGIQDNYT FAQPGIQKKV KALEELVSRI DEQVHNHFRR YEVEYLQFAF RWMNNLLMRE LPLRCTIRLW DTYQSEPEGF SHFHLYVCAA FLIKWRKEIL DEEDFQGLLM LLQNLPTIHW GNEEIGLLLA EAYRLKYMFA DAPNHYRR
Videos