missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TBC1D22B Protein
A cDNA sequence encoding the TBC1D22B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13694-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TBC1D22B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAENSK QFWKRSAKLP GSIQPVYGAQ HPPLDPRLTK NFIKERSKVN TVPLKNKKAS SFHEFARNTS DAWDIGDDEE EDFSSPSFQT LNSKVALATA AQVLENHSKL RVKPERSQST TSDVPANYKV IKSSSDAQLS RNSSDTCLRN PLHKQQSLPL RPIIPLVARI SDQNASGAPP MTVREKTRLE KFRQLLSSQN TDLDELRKCS WPGVPREVRP ITWRLLSGYL PANTERRKLT LQRKREEYFG FIEQYYDSRN EEHHQDTYRQ IHIDIPRTNP LIPLFQQPLV QEIFERILFI WAIRHPASGY VQGINDLVTP FFVVFLSEYV EEDVENFDVT NLSQDMLRSI EADSFWCMSK LLDGIQDNYT FAQPGIQKKV KALEELVSRI DEQVHNHFRR YEVEYLQFAF RWMNNLLMRE LPLRCTIRLW DTYQSEPEGF SHFHLYVCAA FLIKWRKEIL DEEDFQGLLM LLQNLPTIHW GNEEIGLLLA EAYRLKYMFA DAPNHYRR | |
Greater than 80% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TBC1D22B protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4) 30% glycerol and 1mM DTT. |