missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Talin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Talin-1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8498-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054951

  • 726.00€ / 200 microgramos


Fecha estimada de envío
26-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_006280.3
Talin-1 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
7094
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TLN1
Recombinant Protein
E. coli
MLDGTVKTIM VDDSKTVTDM LMTICARIGI TNHDEYSLVR ELMEEKKEEI TGTLRKDKTL LRDEKKMEKL KQKLHTDDEL NWLDHGRTLR EQGVEEHETL LLRRKFFYSD QNVDSRDPVQ LNLLYVQARD DILNGSHPVS FDKACEFAGF QCQIQFGPHN EQKHKAGFLD LKDFLPKEYV KQKGERKIFQ AHKNCGQMSE IEAKVRYVKL ARSLKTYGVS FFLVKEKMKG KNKLVPRLLG ITKECVMRVD EKTKEVIQEW NLTNIKRWAA SPKSFTLDFG DYQDGYYSVQ TTEGEQIAQL IAGYIDII
Videos
Documentos