missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TAF5L Protein
A DNA sequence encoding the Homo sapiens (Human) TAF5L, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6759-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001020418.1 | |
TAF5L Protein | |
Research Use Only | |
TAF5L | |
Recombinant Protein | |
E. coli | |
MKRVRTEQIQ MAVSCYLKRR QYVDSDGPLK QGLRLSQTAE EMAANLTVQS ESGCANIVSA APCQAEPQQY EVQFGRLRNF LTDSDSQHSH EVMPLLYPLF VYLHLNLVQN SPKSTVESFY SRFHGMFLQN ASQKDVIEQL QTTQTIQDIL SNFKLRAFLD NKYVVRLQED SYNYLIRYLQ SDNNTALCKV LTLHIHLDVQ PAKRTDYQLY ASGSSSRSEN NGLEPPDMPS PILQNEAALE VLQESIKRVK DGPPSLTTIC FYAFYNTEQL LNTAEISPDS KLLAAGFDNS CIKLWSLRSK KLKSEPHQVD VSRIHLACDI LEEEV | |
Greater than 80% as determined by SDS-PAGE. |
27097 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |