missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TAF10 Protein

A cDNA sequence encoding the TAF10 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13684-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15924379

  • 103.00€ / 2 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

TAF10 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TAF10 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSGAAPPEG AISNGVYVLP SAANGDVKPV VSSTPLVDFL MQLEDYTPTI PDAVTGYYLN RAGFEASDPR IIRLISLAAQ KFISDIANDA LQHCKMKGTA SGSSRSKSKD RKYTLTMEDL TPALSEYGIN VKKPHYFT
Videos