missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TACO1 Protein
A cDNA sequence encoding the TACO1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13682-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TACO1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSNKWSKVR HIKGPKDVER SRIFSKLCLN IRLAVKEGGP NPEHNSNLAN ILEVCRSKHM PKSTIETALK MEKSKDTYLL YEGRGPGGSS LLIEALSNSS HKCQADIRHI LNKNGGVMAV GARHSFDKKG VIVVEVEDRE KKAVNLERAL EMAIEAGAEDVKETEDEEER NVFKFICDAS SLHQVRKKLD SLGLCSVSCA LEFIPNSKVQ LAEPDLEQAA HLIQALSNHE DVIHVYDNIE | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TACO1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |