missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Survivin / BIRC5 / API4 Protein

A DNA sequence encoding the Homo sapiens (Human) Survivin / BIRC5 / API4, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5719-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937249

  • 675.00€ / 200 microgramos


Fecha estimada de envío
06-07-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001012270.1
Survivin / BIRC5 / API4 Protein
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
332
Greater than 80% as determined by SDS-PAGE.
Research Use Only
BIRC5
Recombinant Protein
E. coli
MGAPTLPPAW QPFLkDaHRI STFKNWPFLE GCACTPERMA EAGFIHCPTE NEPDLAQCFF CFKELEGWEP DDDPIEEHKK HSSGCAFLSV KKQFEELTLG EFLKLDRERA KNKIAKETNN KKKEFEETAK KVRRAIEQLA AMD
Videos
Documentos