missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SURF2 Protein
A cDNA sequence encoding the SURF2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13652-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SURF2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSELPGD VRAFLREHPS LRLQTDARKV RCILTGHELP CRLPELQVYT RGKKYQRLVR ASPAFDYAEF EPHIVPSTKN PHQLFCKLTL RHINKCPEHV LRHTQGRRYQ RALCKYEECQ KQGVEYVPAC LVHRRRRRED QMDGDGPRPR EAFWEPTSSD EGGAASDDSM TDLYPPELFT RKDLGSTEDG DGTDDFLTDK EDEKAKPPRE KATDEGRRET TVYRGLVQKR GKKQLGSLKK KFKSHHRKPK SFSSCKQPG | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SURF2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |