missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SULT1C2 Protein
A cDNA sequence encoding the SULT1C2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13643-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SULT1C2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MALTSDLGKQ IKLKEVEGTL LQPATVDNWS QIQSFEAKPD DLLICTYPKA GTTWIQEIVD MIEQNGDVEK CQRAIIQHRH PFIEWARPPQ PSGVEKAKAM PSPRILKTHL STQLLPPSFW ENNCKFLYVA RNAKDCMVSY YHFQRMNHML PDPGTWEEYF ETFINGKVVW GSWFDHVKGW WEMKDRHQIL FLFYEDIKRD PKHEIRKVMQ FMGKKVDETV LDKIVQETSF EKMKENPMTN RSTVSKSILD QSISSFMRKG TVGDWKNHFT VAQNERFDEI YRRKMEGTSI NFCMEL | |
Greater than 90.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SULT1C2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |