missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Steroid 17-alpha-hydroxylase / 17,20 lyase Protein

A DNA sequence encoding the Homo sapiens (Human) Steroid 17-alpha-hydroxylase/17,20 lyase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5904-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928569

  • 234.00€ / 50 microgramos


Fecha estimada de envío
18-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000093.1
Steroid 17-alpha-hydroxylase / 17,20 lyase Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
1586
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CYP17A1
Recombinant Protein
E. coli
MWELVALLLL TLAYLFWPKR RCPGAKYPKS LLSLPLVGSL PFLPRHGHMH NNFFKLQKKY GPIYSVRMGT KTTVIVGHHQ LAKEVLIKKG KDFSGRPQMA TLDIASNNRK GIAFADSGAH WQLHRRLAMA TFALFKDGDQ KLEKIICQEI STLCDMLATH NGQSIDISFP VFVAVTNVIS LICFNTSYKN GDPELNVIQN YNEGIIDNLS KDSLVDLVPW LKIFPNKTLE KLKSHVKIRN DLLNKILENY KEKFRSDSIT NMLDTLMQAK MNSDNGNAGP DQDSELLSDN HILTTIGDIF GAGVETTTSV VKWTLAFLLH NPQVKKKLYE EIDQNVGFSR TPTISDRNRL LLLEATIREV LRLRPVAPML IPHKANVDSS IGEFAVDKGT EVIINLWALH HNEKEWHQPD QFMPERFLNP AGTQLISPSV SYLPFGAGPR SCIGEILARQ ELFLIMAWLL QRFDLEVPDD GQLPSLEGIP KVVFLIDSFK VKIKVRQAWR EAQAEGST
Videos
Documentos