missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STC 2 Protein
A cDNA sequence encoding the STC 2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13613-HIS-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
8614 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
TDATNPPEGP QDRSSQQKGR LSLQNTAEIQ HCLVNAGDVG CGVFECFENN SCEIRGLHGI CMTFLHNAGKFDAQGKSFIK DALKCKAHAL RHRFGCISRK CPAIREMVSQ LQRECYLKHD LCAAAQENTR VIVEMIHFKDLLLHEPYVDL VNLLLTCGEE VKEAITHSVQ VQCEQNWGSL CSILSFCTSA IQKPPTAPPE RQPQVDRTKLSRAHHGEAGH HLPEPSSRET GRGAKGERGS KSHPNAHARG RVGGLGAQGP SGSSEWEDEQ SEYSDIRRAAADYKDDDDK | |
Greater than 90.0% as determined by SDS-PAGE. |
STC 2 Protein | |
Research Use Only | |
STC2 | |
Human | |
His | |
Filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM Tris buffer, 20mM NaCl, pH 7.5. |