missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SSSCA1 Protein
A cDNA sequence encoding the SSSCA1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13597-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
10534 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMALNGAE VDDFSWEPPT EAETKVLQAR RERQDRISRL MGDYLLRGYR MLGETCADCG TILLQDKQRK IYCVACQELD SDVDKDNPAL NAQAALSQAR EHQLASASEL PLGSRPAPQP PVPRPEHCEG AAAGLKAAQG PPAPAVPPNT DVMACTQTAL LQKLTWASAE LGSSTSLETS IQLCGLIRAC AEALRSLQQL QH | |
Greater than 90.0% as determined by SDS-PAGE. |
SSSCA1 Protein | |
Research Use Only | |
SSSCA1 | |
Human | |
His | |
SSSCA1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |