missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Squalene synthase Protein

A DNA sequence encoding the Homo sapiens (Human) Squalene synthase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6031-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909499

  • 726.00€ / 200 microgramos


Fecha estimada de envío
16-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001274671.1
Squalene synthase Protein
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
2222
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FDFT1
Recombinant Protein
E. coli
EFVKCLGHPE EFYNLVRFRI GGKRKVMPKM DQDSLSSSLK TCYKYLNQTS RSFAAVIQAL DGEMRNAVCI FYLVLRALDT LEDDMTISVE KKVPLLHNFH SFLYQPDWRF MESKEKDRQV LEDFPTISLE FRNLAEKYQT VIADICRRMG IGMAEFLDKH VTSEQEWDKY CHYVAGLVGI GLSRLFSASE FEDPLVGEDT ERANSMGLFL QKTNIIRDYL EDQQGGREFW PQEVWSRYVK KLGDFAKPEN IDLAVQCLNE LITNALHHIP DVITYLSRLR NQSVFNFCAI PQVMAIATLA ACYNNQQVFK GAVKIRKGQA VTLMMDATNM PAVKAIIYQY MEEIYHRIPD SDPSSSKTRQ IISTIRTQNL PNCQLISRSH YSPIYLSFVM LLAALSWQYL TTLSQVTEDY VQTGEH
Videos
Documentos