missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SQSTM1 Protein

A cDNA sequence encoding the SQSTM1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13588-25ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914069

  • 160.00€ / 25 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

SQSTM1 Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SQSTM1 Human solution containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MAMSYVKDDI FRIYIKEKKE CRRDHRPPCA QEAPRNMVHP NVICDGCNGP VVGTRYKCSV CPDYDLCSVC EGKGLHRGHT KLAFPSPFGH LSEGFSHSRW LRKVKHGHFG WPGWEMGPPG NWSPRPPRAG EARPGPTAES ASGPSEDPSV NFLKNVGESV AAALSPLGIE VDIDVEHGGK RSRLTPVSPE SSSTEEKSSS QPSSCCSDPS KPGGNVEGAT QSLAEQMRKI ALESEGRPEE QMESDNCSGG DDDWTHLSSK EVDPSTGELQ SLQMPESEGP SSLDPSQEGP TGLKEAALYP HLPPEADPRL IESLSQMLSM GFSDEGGWLT RLLQTKNYDI GAALDTIQYS KHPPPLLEHH HHHH
Videos