missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPSB2 Protein
A cDNA sequence encoding the SPSB2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13587-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SPSB2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMGQTAL AGGSSSTPTP QALYPDLSCP EGLEELLSAP PPDLGAQRRH GWNPKDCSEN IEVKEGGLYF ERRPVAQSTD GARGKRGYSR GLHAWEISWP LEQRGTHAVV GVATALAPLQ TDHYAALLGS NSESWGWDIG RGKLYHQSKG PGAPQYPAGT QGEQLEVPER LLVVLDMEEG TLGYAIGGTY LGPAFRGLKG RTLYPAVSAV WGQCQVRIRY LGERRAEPHS LLHLSRLCVR HNLGDTRLGQ VSALPLPPAM KRYLLYQ | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SPSB2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |