missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPRY4 Protein
A cDNA sequence encoding the SPRY4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13585-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SPRY4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEPPIPQ SAPLTPNSVM VQPLLDSRMS HSRLQHPLTI LPIDQVKTSH VENDYIDNPS LALTTGPKRT RGGAPELAPT PARCDQDVTH HWISFSGRPS SVSSSSSTSS DQRLLDHMAP PPVADQASPR AVRIQPKVVH CQPLDLKGPA VPPELDKHFL LCEACGKCKC KECASPRTLP SCWVCNQECL CSAQTLVNYG TCMCLVQGIF YHCTNEDDEG SCADHPCSCS RSNCCARWSF MGALSVVLPC LLCYLPATGC VKLAQRGYDR LRRPGCRCKH TNSVICKAAS GDAKTSRPDK PF | |
Greater than 85.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SPRY4 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |